Organic reference standard, Aflatoxin B1-13C17 solution, analytical standard, Supelco®
Catalog # 32764-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:0.5 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:1217449-45-0
- Description:Aflatoxin B₁-¹³C₁₇ solution
- Formula:¹³C₁₇H₁₂O₆
- Storage temperature:−20 °C
- Description:Aflatoxin B₁-¹³C₁₇ solution
- Flash point:2 °C
Specifications
About this item
Aflatoxin B₁-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.
It may be used as an analytical standard for the determination of aflatoxin B₁ in food grade gums, Pu-erh tea samples, animal feed as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).