Organic reference standard, 4-Methyl-3-penten-2-one, analytical standard, Supelco®
Catalog # 49722-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:141-79-7
- Description:4-Methyl-3-penten-2-one, ≥99,0% (GC)
- Melting point:–41,5 °C (lit.)
- Formula:C₆H₁₀O
- Storage temperature:2...8 °C
- Description:4-Methyl-3-penten-2-one
- Flash point:31 °C
- Boiling point:130 °C (lit.)
- Form:Neat
Specifications
About this item
4-Methyl-3-penten-2-one belongs to the volatile organic compounds (VOCs) class of chemicals.
It may be used as a reference standard for the determination of 4-methyl-3-penten-2-one in Irish beef meats by gas chromatography-olfactometry (GC-O), in human urine by static headspace extraction followed by analysis using GC with mass spectrometry (MS), in caja-umbu (Spondias sp.) fruits by GC-MS and in wild gilthead sea bream (Sparus aurata) by simultaneous distillation-extraction (SDE) followed by analysis using GC with flame ionization detector (FID) as well as GC-MS.