Organic reference standard, 4-Hydroxy-2,5-dimethyl-3(2H)-furanone, analytical standard, Supelco®
44869-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:3658-77-3
- Description:4-Hydroxy-2,5-dimethyl-3(2H)-furanone, ≥99,0% (GC)
- Melting point:73...77 °C (lit.)
- Formula:C₆H₈O₃
- Storage temperature:2...8 °C
- Description:4-Hydroxy-2,5-dimethyl-3(2H)-furanone
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.
: Moisture sensitive.