Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 4-Ethylguaiacol, analytical standard, Supelco®
Organic reference standard, 4-Ethylguaiacol, analytical standard, Supelco®
Catalog # 39774-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 4-Ethylguaiacol, analytical standard, Supelco®
Catalog # 39774-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    2785-89-9
  • Description:
    4-Ethylguaiacol, ≥98,0% (GC)
  • Seq:
    01
  • Melting point:
    15 °C (lit.)
  • Formula:
    C₉H₁₂O
  • Storage temperature:
    2...8 °C
  • Description:
    4-Ethylguaiacol
  • Boiling point:
    234...236 °C (lit.)
  • Form:
    Neat

Specifications

About this item