Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, 4-Deoxy-L-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
  06932-50MG
 :  Merck
 :  
Organic reference standard, 4-Deoxy-L-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
  06932-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Description:
    4-Deoxy-L-erythronic acid sodium salt hydrate, ≥97,0% (GC)
  • Seq:
    02
  • Formula:
    C₄H₈O₄ ·xNa+ ·yH₂O
  • Storage temperature:
    2...8 °C
  • Description:
    4-Deoxy-L-erythronic acid sodium salt hydrate

 

 

4-Deoxyerythronic acid has been associated with uremia and diabetes mellitus type 1, but is also a normal organic acid present in human biofluids, derived presumably from L-threonine.