Organic reference standard, 4(5)-Methylimidazole, analytical standard, Supelco®
04535-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:822-36-6
- Description:4(5)-Methylimidazole, ≥98,0% (GC)
- Melting point:44...47 °C (lit.)
- Formula:C₄H₆N₂
- Description:4(5)-Methylimidazole
- Flash point:157 °C
- Boiling point:263 °C (lit.)