Organic reference standard, 3-Methyl-1-butanol, analytical standard, Supelco®
Supplier: Merck
Certificates
About this item
3-Methyl-1-butanol is a potential fuel additive, which is a natural product of fungus and yeast fermentation.
- Utilized as a precursor for the synthesis of various chemicals, such as isoamyl acetate
It may be used as a reference standard for the analysis of 3-methyl-1-butanol content in honey samples using gas chromatography coupled with mass spectrometry (GC-MS).
Specifications
- CAS No.:123-51-3
- Melting point:–117 °C (lit.)
- Formula:C₅H₁₂O
- Description:3-Methyl-1-butanol
- Boiling point:130 °C (lit.)
Specifications
Frequently Bought Together