Organic reference standard, 3-Heptanone, analytical standard, Supelco®
90046-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:106-35-4
- Description:3-Heptanone, ≥98,5% (GC)
- Melting point:–39 °C (lit.)
- Formula:C₇H₁₄O
- Description:3-Heptanone
- Flash point:38 °C
- Boiling point:146...149 °C (lit.)
- Form:Neat