Organic reference standard, 2-Picoline, analytical standard, Supelco®
Catalog # 74350-500MG
Supplier: Merck
CAS Number:
Specifications
- Pk:500 mg
- Standards:Analytical standard
- CAS No.:109-06-8
- Description:2-Picoline, ≥98,5% (GC)
- Melting point:–70 °C (lit.)
- Formula:C₆H₇N
- Description:2-Picoline
- Flash point:29 °C
- Boiling point:128...129 °C (lit.)
- Form:Neat