Organic reference standard, 2-Methylthiophene, analytical standard, Supelco®
78833-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:554-14-3
- Description:2-Methylthiophene, ≥98,5% (GC)
- Seq:01
- Melting point:–63 °C (lit.)
- Formula:C₅H₆S
- Description:2-Methylthiophene
- Flash point:15 °C
- Boiling point:113 °C (lit.)
- Form:Neat