Organic reference standard, 2-Methylbutyraldehyde, analytical standard, Supelco®
Catalog # 61904-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:96-17-3
- Description:2-Methylbutyraldehyde, ≥95,0% (GC)
- Formula:C₅H₁₀O
- Storage temperature:2...8 °C
- Description:2-Methylbutyraldehyde
- Flash point:4 °C
- Boiling point:90...92 °C (lit.)
- Form:Neat