Organic reference standard, 2-Hexanol, analytical standard, Supelco®
Catalog # 41513-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:626-93-7
- Description:2-Hexanol, ≥98,0% (GC)
- Formula:C₆H₁₄O
- Description:2-Hexanol
- Boiling point:136 °C (lit.)