Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,4-Lutidine, analytical standard, Supelco®
Organic reference standard, 2,4-Lutidine, analytical standard, Supelco®
Catalog # 68223-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2,4-Lutidine, analytical standard, Supelco®
Catalog # 68223-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    108-47-4
  • Description:
    2,4-Lutidine, ≥98,5% (GC)
  • Melting point:
    –60 °C (lit.)
  • Formula:
    C₇H₉N
  • Description:
    2,4-Lutidine
  • Boiling point:
    159 °C (lit.)

Specifications

About this item