Organic reference standard, 2,3-Butanediol, analytical standard, Supelco®
42038-250MG
: Merck
:
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:513-85-9
- Description:2,3-Butanediol, ≥97,0% (GC)
- Melting point:25 °C (lit.)
- Formula:C₄H₁₀O₂
- Description:2,3-Butanediol
- Flash point:85 °C
- Boiling point:183...184 °C (lit.)
- Form:Neat
2,3-Butanediol is a chiral compound which is extensively utilised in chemical feedstocks and liquid fuels.
2,3-Butanediol can be used as a reference standard for the determination of 2,3-butanediol in:
'Fino' sherry wine by rotatory and continuous liquid-liquid extraction (LLE) and headspace solid-phase microextraction (HS-SPME) combined with gas chromatography-mass spectrometry (GC-MS). Cell cultures as well as fermentation broths by high performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD)
Cocoa (Theobroma cacao L.) by HS-SPME-GC-MS with principal component analysis (PCA)
Vinegar samples by HS-SPME combined with GC-MS in electron impact ionization (EI+) mode