Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 2,3,5,6-Tetramethylpyrazine, analytical standard, Supelco®
Organic reference standard, 2,3,5,6-Tetramethylpyrazine, analytical standard, Supelco®
Catalog # 95162-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2,3,5,6-Tetramethylpyrazine, analytical standard, Supelco®
Catalog # 95162-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1124-11-4
  • Description:
    2,3,5,6-Tetramethylpyrazine, ≥98,0% (GC)
  • Melting point:
    77...80 °C (lit.)
    85...87 °C
  • Formula:
    C₈H₁₂N₂
  • Description:
    2,3,5,6-Tetramethylpyrazine
  • Boiling point:
    190 °C (lit.)
  • Form:
    Neat

Specifications

About this item