Organic reference standard, 1-Penten-3-ol, analytical standard, Supelco®
01984-250MG
: Merck
:
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:616-25-1
- Description:1-Penten-3-ol, ≥98,0% (GC)
- Formula:C₅H₁₀O
- Description:1-Penten-3-ol
- Flash point:28 °C
- Boiling point:114...115 °C (lit.)