Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 1-Hexanethiol, analytical standard, Supelco®
Organic reference standard, 1-Hexanethiol, analytical standard, Supelco®
Catalog # 75018-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 1-Hexanethiol, analytical standard, Supelco®
Catalog # 75018-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    111-31-9
  • Description:
    1-Hexanethiol, ≥98,0% (GC)
  • Melting point:
    –81...–80 °C (lit.)
  • Formula:
    CH₃(CH₂)₅SH
  • Description:
    1-Hexanethiol
  • Flash point:
    30 °C
  • Boiling point:
    150...154 °C (lit.)

Specifications

About this item