Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, 1-Deoxy-D-ribulose solution, ≥95% (HPLC), Supelco®
Organic reference standard, 1-Deoxy-D-ribulose solution, ≥95% (HPLC), Supelco®
Catalog # 90869-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 1-Deoxy-D-ribulose solution, ≥95% (HPLC), Supelco®
Catalog # 90869-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    ~1 M in H₂O
  • Pk:
    10 mg
  • Description:
    1-Deoxy-D-ribulose solution, ≥95% (HPLC)
  • Seq:
    01
  • Formula:
    C₅H₁₀O₄
  • Storage temperature:
    2...8 °C
  • Description:
    1-Deoxy-D-ribulose solution
  • Form:
    Liquid

Specifications

About this item