Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, 1,4-Dimethoxybenzene, analytical standard, Supelco®
  54306-100MG
 :  Merck
 :  
undefined
Organic reference standard, 1,4-Dimethoxybenzene, analytical standard, Supelco®
  54306-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    150-78-7
  • Description:
    1,4-Dimethoxybenzene, ≥99,0% (GC)
  • Melting point:
    54...56 °C (lit.)
  • Formula:
    C₈H₁₀O₂  
  • Description:
    1,4-Dimethoxybenzene
  • Boiling point:
    213 °C (lit.)