Organic reference standard, (+/-)-Octopamine hydrochloride, analytical standard, Supelco®
68631-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:770-05-8
- Description:(+/-)-Octopamine hydrochloride, ≥98,0% (GC)
- Melting point:~170 °C (dec.) (lit.)
- Formula:C₈H₁₂ClNO₂
- Storage temperature:2…8 °C
- Description:(+/-)-Octopamine hydrochloride
- Form:Neat