Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, (+/-)-1-Amino-2-propanol, analytical standard, Supelco®
  95343-500MG
 :  Merck
 :  
undefined
Organic reference standard, (+/-)-1-Amino-2-propanol, analytical standard, Supelco®
  95343-500MG
 :  Merck
 :  

 

  • Pk:
    500 mg
  • Standards:
    Analytical standard
  • CAS No.:
    78-96-6
  • Description:
    (+/-)-1-Amino-2-propanol, ≥98,0% (GC)
  • Melting point:
    −2 °C (lit.)
  • Formula:
    C₃H₉NO 
  • Storage temperature:
    2…8 °C
  • Description:
    (+/-)-1-Amino-2-propanol
  • Boiling point:
    160 °C (lit.)
  • Form:
    Neat