Organic reference standard, (-)-Caryophyllene oxide, analytical standard, Supelco®
Catalog # 91034-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:1139-30-6
- Description:(-)-Caryophyllene oxide, ≥99,0% (GC)
- Melting point:62...63 °C (lit.)
- Formula:C₁₅H₂₄O
- Storage temperature:2…8 °C
- Description:(-)-Caryophyllene oxide
- Form:Neat