Organic reference standard, (-)-beta-Pinene, analytical standard, Supelco®
Catalog # 80609-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:18172-67-3
- Description:(-)-beta-Pinene, ≥98,5% (sum of enantiomers, GC)
- Formula:C₁₀H₁₆
- Storage temperature:2…8 °C
- Description:(-)-beta-Pinene
- Boiling point:164...165 °C(lit.)
- Form:Neat
Specifications
About this item
(-)-β-Pinene is a monoterpenoid, naturally found in the resins of pine trees and other conifers. It comprises of an exocyclic double bond in its structure.