Order Entry
Ireland
ContactUsLinkComponent
Banded earplugs, X-cap
Banded earplugs, X-cap
Catalog # UVEX2125.361
Supplier:  UVEX
undefined
Banded earplugs, X-cap
Catalog # UVEX2125.361
Supplier:  UVEX

Specifications

  • Description:
    X-cap banded earplugs, SNR 24 dB
  • Pk:
    15

Specifications

About this item

These soft, innovative and unique earplugs ensure a high degree of comfort and stable fit.

  • Ergonomically shaped earplugs for optimal wearer comfort
  • Colour: grey, sky blue, lime
  • 24 decibels insulation - H: 27 dB, M: 19 dB, L: 18 dB
  • Replacement earplugs for hygienic, cost-efficient use

This banded ear protection is ergonomically formed and reduces the transmission of annoying noises by preventing the band from rubbing against clothing.

Certifications: Meets standard EN 352-2.
Packaging: Individually packed in a polythene bag.