
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (by HPLC) mitochondria dye, suitable for fluorescence
Catalog # 70017.
Supplier: Biotium
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:500.93 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:115532-50-8
Formula:C₂₅H₂₅N₂O₃·ClO₄
MDL Number:MFCD00274460