Supelco® Organic Reference Standard, Alkane Standard Mixture for performance tests of GC-systems
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331
Specifications
- Storage conditions:2...30 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Density:0,684 g/ml at 20 °C
- Flash point:–4 °C – Closed cup
Specifications
Frequently Bought Together