Trimethylphenylammonium hydroxide solution ∼0,5 M in methanol, LiChropur™ for GC derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1899-02-1
:
Formula:C₉H₁₅NO
MDL Number:MFCD00041899
:M-Clarity™ quality level = MQ100
:153.23 g/mol
Frequently Bought Together