Order Entry
Ireland
ContactUsLinkComponent
O-Ethylhydroxylamine hydrochloride 99.0-101.0% (by argentometric titration), LiChropur™ for GC derivatisation, Supelco®
O-Ethylhydroxylamine hydrochloride 99.0-101.0% (by argentometric titration), LiChropur™ for GC derivatisation, Supelco®
Catalog # 43504-10G
Supplier:  Merck
CAS Number:  
undefined
O-Ethylhydroxylamine hydrochloride 99.0-101.0% (by argentometric titration), LiChropur™ for GC derivatisation, Supelco®
Catalog # 43504-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Ethoxyamine hydrochloride
 :
 :97.54 g/mol
CAS Number:3332-29-4
Formula:C₂H₈ClNO
MDL Number:MFCD00012956