Order Entry
Ireland
ContactUsLinkComponent
2,3-Dihydroxy-biphenyl ≥98.0% (by HPLC), LiChropur™ for GC derivatisation, Supelco®
2,3-Dihydroxy-biphenyl ≥98.0% (by HPLC), LiChropur™ for GC derivatisation, Supelco®
Catalog # 17403-100MG
Supplier:  Merck
CAS Number:  
undefined
2,3-Dihydroxy-biphenyl ≥98.0% (by HPLC), LiChropur™ for GC derivatisation, Supelco®
Catalog # 17403-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :186.21 g/mol
Melting Pt:108…113 C
 :M-Clarity™ quality level = MQ100
CAS Number:1133-63-7
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00155166