Pesticide standard, Chloridazon-desphenyl, reference material, Honeywell Fluka™
Catalog # HONCR1697-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical Standard
- CAS No.:6339-19-1
- Description:Chloridazon-desphenyl
- Formula:C₄H₄ClN₃O
- Storage temperature:Ambient
- Description:Chloridazon-desphenyl
- Form:Solid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.