Foam swabs
Catalog # TSCLTS/15-C
Supplier: TSC LIMITED
Specifications
- Sterile:+
- Colour:Blue
- Description:Dry foam jumbo swab
- Length:300 mm
- Seq:03
- Shaft/head:PP/foam
- Pk:100
Specifications
About this item
Foam swab is supplied sterile in a tamper evident sealed tube with a write on label or in a peelable pouch.
- Polypropylene (flexible) shaft
- Gamma irradiation to achieve a SAL of 10⁻⁶
- Has a shelf life of two years
This swab is ideal for tricky or hard to reach sampling and also for sampling down small pipes or curved drains.
Caution: These swabs are not suitable for solvents.