Organic reference standard, Zinc oxide, analytical standard, Supelco®
93632-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:1314-13-2
- Description:Zinc oxide
- Formula:ZnO
- Description:Zinc oxide
Zinc oxide has been used as a raw material in the synthesis of environmental friendly silver front contact paste comprising of silver particles, glass frit powders, and resin binder, which can be used in crystalline silicon solar cells.
Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.
: Bottomless glass bottle. Contents are inside inserted fused cone.