Supelco® PESTANAL® Organic Reference Standard, Triforin
45701-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:26644-46-2
- Description:Triforin
- Storage conditions:2...30 °C
- Hazard class description:H332
- Shipping conditions:STD
- MDL No.:MFCD00055521
- M-Clarity quality level:MQ100
- Molecular weight:434,96
- GHS signal: GHS07
: P261 - P271 - P304 + P340 + P312