Supelco® PESTANAL® Organic Reference Standard, Triazamate
68723-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:112143-82-5
- Description:Triazamate
- Storage conditions:2...30 °C
- Hazard class description:H301 - H319 - H330 - H400
- Shipping conditions:STD
- MDL No.:MFCD01632354
- M-Clarity quality level:MQ100
- Synonym(s):Ethyl (3-tert-butyl-1-dimethylcarbamoyl-1H-1,2,4-triazol-5-ylthio)acetate; Ethyl 2-{{1-[(dimethylamino)carbonyl]-3-(1,1-dimethylethyl)-1H-1,2,4-triazol-5-yl}thio}acetate
- Melting point:52 - 56 °C
- Molecular weight:314,40
- Flash point:189 °C
- GHS signal: GHS06, GHS09
: P273 - P301 + P310 + P330 - P304 + P340 + P310 - P305 + P351 + P338