Pesticide reference standard, Tralomethrin, PESTANAL®, analytical standard, Supelco®
Catalog # 32531-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- Description:Tralomethrin
- Formula:C₂₂H₁₉Br₄NO₃
- Storage temperature:2...8 °C
- Description:Tralomethrin