Supelco® PESTANAL® Organic Reference Standard, Thifluzamide
49792-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:130000-40-7
- Description:Thifluzamide
- Storage conditions:2…8 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD03428071
- M-Clarity quality level:MQ100
- Synonym(s):2′,6′-Dibromo-2-methyl-4′-trifluoromethoxy-4-trifluoromethyl-1,3-thiazole-5-carboxanilide; N-[2,6-Dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-5-thiazolecarboxamide
- Molecular weight:528,06
- GHS signal: GHS09
: P273 - P391 - P501