Supelco® PESTANAL® Organic Reference Standard, Terbumeton
Catalog # 31527-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:33693-04-8
- Description:Terbumeton
- Storage conditions:2...30 °C
- Hazard class description:H302 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055297
- M-Clarity quality level:MQ100
- Formula:C₁₀H₁₉N₅O
- Molecular weight:225,29
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501