Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330-Cis-Enol
Catalog # 32484-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:203312-38-3
- Description:Spirotetramat Metabolite BYI08330-cis-enol
- Seq:02
- Storage conditions:–20 °C
- Hazard class description:H317 - H319 - H335 - H361fd - H410
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]dec-3-en-2-one
- Formula:C₁₈H₂₃NO₃
- Molecular weight:301,38
- Storage temperature:−20 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS09
Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.
Caution: P202 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P308 + P313