Supelco® PESTANAL® Organic Reference Standard, Resmethrin
45655-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard, mixture of isomers
- CAS No.:10453-86-8
- Description:Resmethrin
- Storage conditions:2...30 °C
- Hazard class description:H302 - H410
- Shipping conditions:STD
- MDL No.:MFCD00041806
- M-Clarity quality level:MQ100
- Formula:C₂₂H₂₆O₃
- Molecular weight:338,44
- Flash point:Not applicable
- GHS signal: GHS07, GHS09
: P264 - P270 - P273 - P301 + P312 - P391 - P501