Pesticide reference standards, Pyridaphenthion, PESTANAL®, analytical standards, Supelco®
32538-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:119-12-0
- Description:Pyridaphenthion
- Formula:C₁₄H₁₇N₂O₄PS
- Description:Pyridaphenthion
Pyridaphenthion may be used as a reference standard for the quantification of pyridaphenthion in biological samples using liquid chromatography-atmospheric pressure chemical ionisation mass spectrometry.
- This insecticide is effective against sucking and chewing insects and spider mites