Supelco® PESTANAL® Organic Reference Standard, Pyraclostrobin
Catalog # 33696-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:175013-18-0
- Description:Pyraclostrobin
- Storage conditions:2...30 °C
- Hazard class description:H315 - H331 - H410
- Shipping conditions:STD
- MDL No.:MFCD03792774
- M-Clarity quality level:MQ100
- Synonym(s):Methyl {2-[1-(4-chlorophenyl)-1H-pyrazol-3-yloxymethyl]phenyl}methoxycarbamate
- Formula:C₁₉H₁₈ClN₃O₄
- Molecular weight:387,82
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P261 - P264 - P271 - P273 - P302 + P352 - P304 + P340 + P311