Supelco® PESTANAL® Organic Reference Standard, Phenmedipham
36192-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:13684-63-4
- Description:Phenmedipham
- Storage conditions:2...30 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD00055419
- M-Clarity quality level:MQ100
- Formula:C₁₆H₁₆N₂O₄
- Molecular weight:300,31
- Flash point:100 °C - closed cup
- GHS signal: GHS09
: P273 - P391 - P501