Order Entry
Ireland
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Penconazol
Catalog # 36189-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Penconazol
Catalog # 36189-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    66246-88-6
  • Description:
    Penconazol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H361d - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078737
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₃H₁₅Cl₂N₃
  • Molecular weight:
    284,18
  • Flash point:
    100 °C - closed cup

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P201 - P273 - P301 + P312 + P330 - P308 + P313