Organic reference standard, Octamethylcyclotetrasiloxane, analytical standard, Supelco®
43883-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:556-67-2
- Description:Octamethylcyclotetrasiloxane
- Melting point:17...18 °C (lit.)
- Formula:C₈H₂₄O₄Si₄
- Description:Octamethylcyclotetrasiloxane
- Boiling point:175...176 °C (lit.)