Order Entry
Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Nitenpyram
Supelco® PESTANAL® Organic Reference Standard, Nitenpyram
Catalog # 46077-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Nitenpyram
Catalog # 46077-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    150824-47-8
  • Description:
    Nitenpyram
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01631161
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    270,72

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P270 - P301 + P312 - P302 + P352 - P305 + P351 + P338