Supelco® PESTANAL® Organic Reference Standard, Metconazole
37909-100MG-R
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard, mixture of stereoisomers
- CAS No.:125116-23-6
- Description:Metconazole
- Storage conditions:2...30 °C
- Hazard class description:H302 - H361d - H411
- Shipping conditions:STD
- MDL No.:MFCD01632341
- M-Clarity quality level:MQ100
- Synonym(s):5-(4-Chlorophenylmethyl)-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)cyclopentanol
- Formula:C₁₇H₂₂ClN₃O
- Molecular weight:319,83
- Flash point:Not applicable
- GHS signal: GHS08, GHS07, GHS09
: P201 - P202 - P264 - P273 - P301 + P312 - P308 + P313