Supelco® PESTANAL® Organic Reference Standard, Mecarbam
Catalog # 36515-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:2595-54-2
- Description:Mecarbam
- Storage conditions:2...8 °C
- Hazard class description:H300 + H330 - H311 - H410
- Shipping conditions:STD
- MDL No.:MFCD00129669
- M-Clarity quality level:MQ100
- Formula:C₁₀H₂₀NO₅PS₂
- Molecular weight:329,37
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310