Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Leucocrystal Violet-d6, analytical standard, Supelco®
Organic reference standard, Leucocrystal Violet-d6, analytical standard, Supelco®
Catalog # 32834-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Leucocrystal Violet-d6, analytical standard, Supelco®
Catalog # 32834-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1173023-92-1
  • Description:
    Leucocrystal Violet-d6
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₅D₆H₂₅N₃
  • Description:
    Leucocrystal Violet-d6

Specifications

About this item

Leucocrystal violet-d₆ might be used as an internal standard in the determination of triphenylmethane dyes in rainbow trout samples using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).