Order Entry
Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Imazapyr
Supelco® PESTANAL® Organic Reference Standard, Imazapyr
Catalog # 37877-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Imazapyr
Catalog # 37877-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    81334-34-1
  • Description:
    Imazapyr
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00144470
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₃H₁₅N₃O₃
  • Molecular weight:
    261,28
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P280 - P305 + P351 + P338 - P337 + P313