Order Entry
Ireland
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Hexaflumuron
Catalog # 37902-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Hexaflumuron
Catalog # 37902-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    86479-06-3
  • Description:
    Hexaflumuron
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01743935
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-[3,5-Dichloro-4-(1,1,2,2-tetrafluoroethoxy)phenylcarbamoyl]-2,6-difluorobenzamide
  • Formula:
    C₁₆H₈Cl₂F₆N₂O₃
  • Molecular weight:
    461,14
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P273 - P391 - P501